Boron trifluoride etherate, approx. 48% BF3
Boron trifluoride etherate, approx. 48% BF3
Boron trifluoride etherate, approx. 48% BF3
Thermo Scientific Chemicals

Boron trifluoride etherate, approx. 48% BF3

CAS: 109-63-7 | C4H10BF3O | 141.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 kg
2.5 kg
Packaging:
Glass bottle
Catalog number 174560250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS109-63-7
IUPAC Nameethoxyethane; trifluoroborane
Molecular FormulaC4H10BF3O
InChI KeyKZMGYPLQYOPHEL-UHFFFAOYSA-N
SMILESFB(F)F.CCOCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Assay47.5 to 49 % (BF3)
Refractive index1.3429 to 1.3449 (20°C, 589 nm)
Appearance (Color)Clear colorless to straw yellow
AppearanceMay darken on storage
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only