Thermanox Cover Slip, 22 x 60 mm Rectangular, TC Surface, Pack of 50 - FAQs
Thermanox Cover Slip, 22 x 60 mm Rectangular, TC Surface, Pack of 50 - FAQs
View additional product information for Nunc™ Thermanox™ Coverslips - FAQs (174934, 174969, 174942, 174985, 174977, 150067, 174950)
9 product FAQs found
Why are Nunc Thermanox Coverslips not recommended for techniques involving fluorescent stains?
Thermanox Coverslips are auto-fluorescent within the 380nm - 545nm range.
What are the dimensions of your Nunc Thermanox Coverslips?
Please review the following information regarding the dimensions of our Nunc Thermanox Coverslips:
Cat. No.; Dimensions:
- 174950; 13 mm diameter
- 174969; 15 mm diameter
- 174977; 22 mm diameter
- 174985; 25 mm diameter
- 150067; 24 mm x 30 mm
- 174934; 10.5 mm x 22 mm
- 174942; 22 mm x 60 mm
What is the thickness rating for your Nunc Thermanox Coverslips?
Our Nunc Thermanox Coverslips are 0.2 mm thickness.
The thickness grade ranges as follows:
- No. 0: 0.085-0.13 mm
- No. 1: 0.13-0.16 mm
- No. 1.5: 0.16-0.19 mm
- No. 2: 0.19-0.23 mm
Based upon the above thickness grade ranges, our Nunc Thermanox Coverslips have a No.2 thickness rating.
What is the thickness of your Nunc Thermanox Coverslips?
Our Nunc Thermanox Coverslips are 0.2 mm in thickness.
Can I autoclave the Nunc Thermanox Coverslips?
Yes, our Nunc Thermanox Coverslips can be autoclaved at 20 min steam, 20 min dry, at 120 degrees C.
When I am using Nunc Thermanox Coverslips for fluorescence, at what range does it exhibit autofluorescence?
Our Nunc Thermanox Coverslips autofluoresce at the rhodamine and FITC range.
Fluorescein isothiocyanate:
- excitation = 494 nm
- emission = 517 nm
How can I determine which side, of your Nunc Thermanox Coverslips, is cell culture-treated?
A simple method, to determine which side of our Nunc Thermanox Coverslip is cell cultured-treated, is the "droplet" test. A drop of water or culture medium will spread on the hydrophilic, cell culture treated side.
Please review the following procedure for the "droplet" test:
- Put a drop of sterile media or water on the coverslip
- If the droplet forms a bead, then that side of the coverslip is not treated
- If the droplet spreads evenly over the coverslip, then that side of the coverslip is treated
Additionally, the coverslips are packaged so the cell culture-treated side faces the label.
What is Thermanox plastic?
Thermanox is a trademark name for a particular type of polymer in the polyolefin family.
How can I tell which side of your Thermanox coverslips is treated?
If Thermanox coverslips are still in the original packaging, the side facing the label is treated. If they have been removed, there is another way to determine this: hold one with forceps at a slight angle to the horizontal and dispense a few drops of sterile water onto the surface. On the untreated side the water will bead up tightly as it runs off, but on the treated side it will spread out much more due to the hydrophilic surface modification.