Triethyloxonium tetrafluoroborate, 1M solution in methylene chloride
Triethyloxonium tetrafluoroborate, 1M solution in methylene chloride
Triethyloxonium tetrafluoroborate, 1M solution in methylene chloride
Thermo Scientific Chemicals

Triethyloxonium tetrafluoroborate, 1M solution in methylene chloride

CAS: 368-39-8 | C6H15BF4O | 189.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Catalog number 175111000
Price (USD)
133.65
Special offer
Online exclusive
Ends: 28-Dec-2026
157.00
Save 23.35
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
133.65
Special offer
Online exclusive
Ends: 28-Dec-2026
157.00
Save 23.35
Each
Chemical Identifiers
CAS368-39-8
IUPAC Nametetrafluoroboranuide; triethyloxidanium
Molecular FormulaC6H15BF4O
InChI KeyIYDQMLLDOVRSJJ-UHFFFAOYSA-N
SMILESF[B-](F)(F)F.CC[O+](CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Appearance (Color)Clear colorless to yellow to light brown
Strength0.9 to 1.1 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only