Nitronium tetrafluoroborate, 0.5M solution in sulfolane
Nitronium tetrafluoroborate, 0.5M solution in sulfolane
Nitronium tetrafluoroborate, 0.5M solution in sulfolane
Nitronium tetrafluoroborate, 0.5M solution in sulfolane
Nitronium tetrafluoroborate, 0.5M solution in sulfolane
Thermo Scientific Chemicals

Nitronium tetrafluoroborate, 0.5M solution in sulfolane

CAS: 13826-86-3 | BF4NO2 | 132.81 g/mol
数量:
25 mL
200 mL
パッケージ:
Glass bottle
製品番号(カタログ番号) 175130250
価格(JPY)
-
数量:
25 mL
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS13826-86-3
IUPAC Namedioxoamine; tetrafluoroboranuide
Molecular FormulaBF4NO2
InChI KeyXYRQURYIJKJVSF-UHFFFAOYSA-N
SMILES[O-][NH+]=O.F[B-](F)(F)F
さらに表示
Strength0.48 to 0.55 M
Appearance (Color)Colorless to yellow
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only