Diethyl ether, 99.5%, for analysis, stabilized with BHT, conforms to Eur.Ph.
Diethyl ether, 99.5%, for analysis, stabilized with BHT, conforms to Eur.Ph.
Diethyl ether, 99.5%, for analysis, stabilized with BHT, conforms to Eur.Ph.
Thermo Scientific Chemicals

Diethyl ether, 99.5%, for analysis, stabilized with BHT, conforms to Eur.Ph.

CAS: 60-29-7 | C4H10O | 74.12 g/mol
数量:
1 L
2.5 L
5 L
25 L
パッケージ:
すべて表示
Glass Bottle
製品番号(カタログ番号) 176820010
価格(JPY)
-
数量:
1 L
パッケージ:
Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS60-29-7
IUPAC Nameethoxyethane
Molecular FormulaC4H10O
InChI KeyRTZKZFJDLAIYFH-UHFFFAOYSA-N
SMILESCCOCC
さらに表示
Nickel (Ni)=<0.02 ppm
Methanol=<0.01 %
Acetone=<0.002 %
Iron (Fe)=<0.1 ppm
Cobalt (Co)=<0.02 ppm
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only