Poly(methyl methacrylate), beads, average M.W. 35,000
Poly(methyl methacrylate), beads, average M.W. 35,000
Poly(methyl methacrylate), beads, average M.W. 35,000
Thermo Scientific Chemicals

Poly(methyl methacrylate), beads, average M.W. 35,000

CAS: 9011-14-7 | (C5H8O2)n | NaN g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
1 kg
Packaging:
View all
Glass bottle
Catalog number 178760250
Price (EUR)
57,65
Online Exclusive
63,50
Save 5,85 (9%)
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
57,65
Online Exclusive
63,50
Save 5,85 (9%)
Each
Chemical Identifiers
CAS9011-14-7
Molecular Formula(C5H8O2)n
SMILESCOC(=O)C(C)(-*)C-*
MDL NumberMFCD00134349
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Beads
Loss on drying=<5 %
Particle size140 to 250 microns (approximately)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only