3,3-Dimethylbutylamine, 95%
3,3-Dimethylbutylamine, 95%
3,3-Dimethylbutylamine, 95%
Thermo Scientific Chemicals

3,3-Dimethylbutylamine, 95%

CAS: 15673-00-4 | C6H15N | 101.19 g/mol
数量:
1 g
5 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 179380050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS15673-00-4
IUPAC Name3,3-dimethylbutan-1-amine
Molecular FormulaC6H15N
InChI KeyGPWHFPWZAPOYNO-UHFFFAOYSA-N
SMILESCC(C)(C)CCN
さらに表示
Appearance (Color)Clear colorless to yellow
GC>=94 %
Infrared spectrumConforms
Refractive index1.4120 to 1.4160 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only