Tetrahydrofuran-D8, for NMR, 99.5 atom % D
Tetrahydrofuran-D8, for NMR, 99.5 atom % D
Tetrahydrofuran-D8, for NMR, 99.5 atom % D
Thermo Scientific Chemicals

Tetrahydrofuran-D8, for NMR, 99.5 atom % D

CAS: 1693-74-9 | C4H8O | 80.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 mL
5 mL
10 mL
25 mL
Packaging:
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 180130010
Price (USD)
-
Quantity:
1 mL
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS1693-74-9
IUPAC Name(²H₈)oxolane
Molecular FormulaC4H8O
InChI KeyWYURNTSHIVDZCO-SVYQBANQSA-N
SMILES[2H]C1([2H])OC([2H])([2H])C([2H])([2H])C1([2H])[2H]
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.06 % (Coulometric)
Appearance (Color)Clear colorless
Appearance (Form)Liquid
D-Enrichment>=99.4 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
RUO – Research Use Only