tert-Butyl hydroperoxide, 70% Solution in water
tert-Butyl hydroperoxide, 70% Solution in water
tert-Butyl hydroperoxide, 70% Solution in water
Thermo Scientific Chemicals

tert-Butyl hydroperoxide, 70% Solution in water

CAS: 75-91-2 | C4H10O2 | 90.12 g/mol
数量:
5 g
250 g
500 g
2.5 kg
10 kg
パッケージ:
すべて表示
Plastic bottle
製品番号(カタログ番号) 180340025
価格(JPY)
-
数量:
2.5 kg
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS75-91-2
IUPAC Name2-methylpropane-2-peroxol
Molecular FormulaC4H10O2
InChI KeyCIHOLLKRGTVIJN-UHFFFAOYSA-N
SMILESCC(C)(C)OO
さらに表示
Color scale=<20 APHA
Appearance (Form)Clear solution
Titration Iodometric68.5 to 71.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only