1,1,2,2-Tetrabromoethane, 98%
1,1,2,2-Tetrabromoethane, 98%
1,1,2,2-Tetrabromoethane, 98%
Thermo Scientific Chemicals

1,1,2,2-Tetrabromoethane, 98%

CAS: 79-27-6 | C2H2Br4 | 345.65 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Catalog number 180870025
Price (USD)
-
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS79-27-6
IUPAC Name1,1,2,2-tetrabromoethane
Molecular FormulaC2H2Br4
InChI KeyQXSZNDIIPUOQMB-UHFFFAOYSA-N
SMILESBrC(Br)C(Br)Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Refractive index1.6355 to 1.6375 (20°C, 589 nm)
Color scale=<75 APHA
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only