Poly(vinyl acetate), approx. M.W. 100,000
Poly(vinyl acetate), approx. M.W. 100,000
Poly(vinyl acetate), approx. M.W. 100,000
Poly(vinyl acetate), approx. M.W. 100,000
Thermo Scientific Chemicals

Poly(vinyl acetate), approx. M.W. 100,000

CAS: 9003-20-7 | (C4H6O2)n | 86.09 g/mol
Have Questions?
Catalog NumberQuantityPackaging
183265000500 gGlass bottle
Catalog number 183265000
Price (USD)
126.65
Special offer
Online exclusive
Ends: 28-Dec-2026
149.00
Save 22.35
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
126.65
Special offer
Online exclusive
Ends: 28-Dec-2026
149.00
Save 22.35
Each
Chemical Identifiers
CAS9003-20-7
Molecular Formula(C4H6O2)n
SMILESCC(=O)OC(-*)C-*
MDL NumberMFCD00084457
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear
Loss on drying=<5 %
Appearance (Form)Beads
Infrared spectrumConforms

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only