2,3-Dihydroxybenzaldehyde, 97%
2,3-Dihydroxybenzaldehyde, 97%
2,3-Dihydroxybenzaldehyde, 97%
Thermo Scientific Chemicals

2,3-Dihydroxybenzaldehyde, 97%

CAS: 24677-78-9 | C7H6O3 | 138.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 183510250
Price (USD)
159.65
Special offer
Online exclusive
Ends: 28-Dec-2026
188.00
Save 28.35
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
159.65
Special offer
Online exclusive
Ends: 28-Dec-2026
188.00
Save 28.35
Each
Chemical Identifiers
CAS24677-78-9
IUPAC Name2,3-dihydroxybenzaldehyde
Molecular FormulaC7H6O3
InChI KeyIXWOUPGDGMCKGT-UHFFFAOYSA-N
SMILESOC1=CC=CC(C=O)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to green-beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with NaOH>=96.0 %
GC>=96.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only