1H-Benzotriazole, 99%
1H-Benzotriazole, 99%
1H-Benzotriazole, 99%
Thermo Scientific Chemicals

1H-Benzotriazole, 99%

CAS: 95-14-7 | C6H5N3 | 119.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 183880025
Price (USD)
-
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS95-14-7
IUPAC Name2H-1,2,3-benzotriazole
Molecular FormulaC6H5N3
InChI KeyQRUDEWIWKLJBPS-UHFFFAOYSA-N
SMILESN1N=C2C=CC=CC2=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to light beige
Appearance (Form)flakes
Melting point94°C to 99°C
Appearance (Form)Powder or crystals or granules or needles or
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only