3,3'-Dimethoxybenzidine dihydrochloride, 98%
3,3'-Dimethoxybenzidine dihydrochloride, 98%
3,3'-Dimethoxybenzidine dihydrochloride, 98%
Thermo Scientific Chemicals

3,3'-Dimethoxybenzidine dihydrochloride, 98%

CAS: 20325-40-0 | C14H16N2O2·2HCl | 317.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 184470250
Price (TWD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS20325-40-0
IUPAC Namedihydrogen 3,3'-dimethoxy-[1,1'-biphenyl]-4,4'-diamine dichloride
Molecular FormulaC14H18Cl2N2O2
InChI KeyUXTIAFYTYOEQHV-UHFFFAOYSA-N
SMILES[H+].[H+].[Cl-].[Cl-].COC1=CC(=CC=C1N)C1=CC=C(N)C(OC)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige to grey to purple-grey
Titration with HClO4>=97.5 % (on dry substance)
Loss on drying=<5 %
Appearance (Form)Powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only