Thermo Scientific Chemicals

N-(2-Chloroethyl)acetamide, 98%, Thermo Scientific Chemicals

Catalog number: 184520500
50 g, Each
Thermo Scientific Chemicals

N-(2-Chloroethyl)acetamide, 98%, Thermo Scientific Chemicals

Catalog number: 184520500
50 g, Each
Quantity
Catalog number: 184520500
Price (USD)
Price: 152.00
Online price: 132.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7355-58-0
IUPAC Name
N-(2-chloroethyl)acetamide
Molecular Formula
C4H8ClNO
InChI Key
HSKNJSHFPPHTAQ-UHFFFAOYSA-N
SMILES
CC(=O)NCCCl
Appearance (Form)
Viscous liquid
Infrared spectrum
Conforms
Refractive index
1.4770 to 1.4800 (20°C, 589 nm)
Appearance (Color)
Clear colorless to light yellow
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text