Hexanoic anhydride, 97%
Hexanoic anhydride, 97%
Hexanoic anhydride, 97%
Thermo Scientific Chemicals

Hexanoic anhydride, 97%

CAS: 2051-49-2 | C12H22O3 | 214.3 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Glass bottle
Catalog number 186750500
Price (EUR)
24,30
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
24,30
Each
Chemical Identifiers
CAS2051-49-2
IUPAC Namehexanoyl hexanoate
Molecular FormulaC12H22O3
InChI KeyPKHMTIRCAFTBDS-UHFFFAOYSA-N
SMILESCCCCCC(=O)OC(=O)CCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4270 to 1.4300 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Titration by coupling>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only