sec-Butyllithium, 1.3M sol. in cyclohexane/hexane (92/8), AcroSeal™, Thermo Scientific Chemicals
sec-Butyllithium, 1.3M sol. in cyclohexane/hexane (92/8), AcroSeal™, Thermo Scientific Chemicals
sec-Butyllithium, 1.3M sol. in cyclohexane/hexane (92/8), AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

sec-Butyllithium, 1.3M sol. in cyclohexane/hexane (92/8), AcroSeal™, Thermo Scientific Chemicals

Catalog NumberQuantityPackaging
187541000100 mLAcroSeal™ Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 187541000
Price (JPY)
-
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS598-30-1
IUPAC Name(butan-2-yl)lithium
Molecular FormulaC4H9Li
InChI KeyVATDYQWILMGLEW-UHFFFAOYNA-N
SMILES[Li]C(C)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear to hazy colorless to yellow
Appearance (Form)Solution
AppearanceMay develop some turbidity or precipitate
Titration Active base10.5 to 12.5 %
Titration Active base1.23 to 1.46 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only