4-O-Methyldopamine hydrochloride, 98%
4-O-Methyldopamine hydrochloride, 98%
4-O-Methyldopamine hydrochloride, 98%
Thermo Scientific Chemicals

4-O-Methyldopamine hydrochloride, 98%

CAS: 645-33-0 | C9H13NO2·HCl | 203.67 g/mol
数量:
50 mg
500 mg
パッケージ:
Glass bottle
製品番号(カタログ番号) 187745000
価格(JPY)
-
数量:
500 mg
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS645-33-0
IUPAC Namehydrogen 5-(2-aminoethyl)-2-methoxyphenol chloride
Molecular FormulaC9H14ClNO2
InChI KeyKAAFITWSSODFMA-UHFFFAOYSA-N
SMILES[H+].[Cl-].COC1=CC=C(CCN)C=C1O
さらに表示
Melting point207°C to 211°C
HPLC>=97.5 %
Appearance (Color)White to Light beige
Appearance (Form)Powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only