Ethyl 4-methyl-5-imidazolecarboxylate, 98%
Ethyl 4-methyl-5-imidazolecarboxylate, 98%
Ethyl 4-methyl-5-imidazolecarboxylate, 98%
Thermo Scientific Chemicals

Ethyl 4-methyl-5-imidazolecarboxylate, 98%

CAS: 51605-32-4 | C7H10N2O2 | 154.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 190340500
Price (USD)
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS51605-32-4
IUPAC Nameethyl 5-methyl-1H-imidazole-4-carboxylate
Molecular FormulaC7H10N2O2
InChI KeyVLDUBDZWWNLZCU-UHFFFAOYSA-N
SMILESCCOC(=O)C1=C(C)NC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
HPLC>=97.5 %
Appearance (Color)Off-white to tan
Infrared spectrumConforms
Melting point204°C to 208°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only