Product Image
Thermo Scientific Chemicals

Aluminum oxide, for chromatography, neutral, Brockmann I, 50-200 μm, 90A

 
製品番号(カタログ番号)数量
1904100101 kg
製品番号(カタログ番号) 190410010
価格(JPY)
-
数量:
1 kg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1344-28-1
IUPAC Namedialuminium(3+) trioxidandiide
Molecular FormulaAl2O3
InChI KeyPNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Al+3].[Al+3]
さらに表示
Appearance (Form)Crystalline powder
Appearance (Color)White
pH6.5 to 7.5
Particle size=<10 % (< 50 um)
Specific surface area90 to 170 m2/g,N2 adsorp
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only