Mercury(II) sulfate, 99+%
Mercury(II) sulfate, 99+%
Mercury(II) sulfate, 99+%
Thermo Scientific Chemicals

Mercury(II) sulfate, 99+%

CAS: 7783-35-9 | HgO4S | 296.65 g/mol
Quantity:
100 g
500 g
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 190501000
Price (JPY)
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS7783-35-9
IUPAC Namemercury(2+) sulfate
Molecular FormulaHgO4S
InChI KeyDOBUSJIVSSJEDA-UHFFFAOYSA-L
SMILES[Hg++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Assay>=99.0 %
Appearance (Color)White to off-white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only