Ethyl cellulose, ethoxyl content 48%, 22 cps
Ethyl cellulose, ethoxyl content 48%, 22 cps
Ethyl cellulose, ethoxyl content 48%, 22 cps
Thermo Scientific Chemicals

Ethyl cellulose, ethoxyl content 48%, 22 cps

CAS: 9004-57-3 | C23H24N6O4 | 448.483 g/mol
Quantity:
250 g
500 g
2.5 kg
Packaging:
View all
Plastic drum
Catalog number 190970025
Price (JPY)
-
Quantity:
2.5 kg
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
CAS9004-57-3
IUPAC Name2-[butyl({4-[2-(2,6-dicyano-4-nitrophenyl)diazen-1-yl]-3-methylphenyl})amino]ethyl acetate
Molecular FormulaC23H24N6O4
InChI KeyARSKJXYLLONUAJ-UHFFFAOYSA-N
SMILESCCCCN(CCOC(C)=O)C1=CC(C)=C(C=C1)N=NC1=C(C=C(C=C1C#N)[N+]([O-])=O)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Viscosity18 to 24 cP (5% soln. in 80/20 toluene/ethanol, 25°C)
Infrared spectrumConforms
Appearance (Form)Crystalline powder or granules
Assay48 to 49.5 % (Ethoxyl)
Appearance (Color)White to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only