Osmium tetroxide, 99.9+%, (trace metal basis)
Osmium tetroxide, 99.9+%, (trace metal basis)
Osmium tetroxide, 99.9+%, (trace metal basis)
Thermo Scientific Chemicals

Osmium tetroxide, 99.9+%, (trace metal basis)

CAS: 20816-12-0 | O4Os | 254.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
250 mg
500 mg
1 g
5 g
Catalog number 191180010
Price (USD)
276.65
Online exclusive
307.00
Save 30.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
276.65
Online exclusive
307.00
Save 30.35
Each
Chemical Identifiers
CAS20816-12-0
IUPAC Nametetraoxoosmium
Molecular FormulaO4Os
InChI KeyVUVGYHUDAICLFK-UHFFFAOYSA-N
SMILESO=[Os](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Osmium (Os)74.2 to 75.4 %
SolubilityPasses test
Appearance (Form)Crystals
Non-volatile matter=<0.2 %
Ruthenium (Ru)=<0.01 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis, Oxidation reagent
RUO – Research Use Only