Lithium hexafluorophosphate, 98%, pure
Lithium hexafluorophosphate, 98%, pure
Lithium hexafluorophosphate, 98%, pure
Lithium hexafluorophosphate, 98%, pure
Thermo Scientific Chemicals

Lithium hexafluorophosphate, 98%, pure

CAS: 21324-40-3 | F6LiP | 151.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Plastic bottle
Catalog number 191260050
Price (USD)
51.00
Each
Quantity:
5 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
51.00
Each
Specifications
Assay Percent Range97.5% min. (ex Li)
CAS21324-40-3
Chemical Name or MaterialLithium hexafluorophosphate
ColorWhite
EINECS Number244-334-7
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only