Calcium chloride, 96%, for biochemistry, anhydrous
Calcium chloride, 96%, for biochemistry, anhydrous
Calcium chloride, 96%, for biochemistry, anhydrous
Thermo Scientific Chemicals

Calcium chloride, 96%, for biochemistry, anhydrous

CAS: 10043-52-4 | CaCl2 | 110.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 192731000
Price (USD)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS10043-52-4
IUPAC Namecalcium dichloride
Molecular FormulaCaCl2
InChI KeyUXVMQQNJUSDDNG-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Ca++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Heavy metals (as Pb)=<5 ppm
Copper (Cu)=<0.0005 %
Phosphate (PO4)=<0.001 %
Sulfate (SO4)=<0.02 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only