Germanium, 99.999%, (trace metal basis), powder
Germanium, 99.999%, (trace metal basis), powder
Germanium, 99.999%, (trace metal basis), powder
Thermo Scientific Chemicals

Germanium, 99.999%, (trace metal basis), powder

CAS: 7440-56-4 | Ge | 72.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 193110500
Price (USD)
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7440-56-4
IUPAC Namegermanium
Molecular FormulaGe
InChI KeyGNPVGFCGXDBREM-UHFFFAOYSA-N
SMILES[Ge]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Glistening crystalline powder
Total trace metal impurities=<0.001 %
Appearance (Color)Grey to black
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only