Lithium chloride, 99%, extra pure
Lithium chloride, 99%, extra pure
Lithium chloride, 99%, extra pure
Thermo Scientific Chemicals

Lithium chloride, 99%, extra pure

CAS: 7447-41-8 | ClLi | 42.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle
Catalog number 193370050
Price (USD)
1,105.00
Each
Quantity:
5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
1,105.00
Each
Chemical Identifiers
CAS7447-41-8
IUPAC Namelithium(1+) chloride
Molecular FormulaClLi
InChI KeyKWGKDLIKAYFUFQ-UHFFFAOYSA-M
SMILES[Li+].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals or granules
Loss on drying=<0.5 % (400°C, 4 h)
Insoluble matter=<0.01 % (in acid)
Titration Argentometric>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Frequently asked questions (FAQs)

What storage conditions should I use for Lithium chloride?

Lithium chloride is hygroscopic and should be stored under a dry, inert atmosphere, and protected from sunlight. Refer to the product label for specific storage temperature requirements.