Ammoniumdihydrogenphosphat, 99.9 %, reinst, Thermo Scientific Chemicals
Ammoniumdihydrogenphosphat, 99.9 %, reinst, Thermo Scientific Chemicals
Ammoniumdihydrogenphosphat, 99.9 %, reinst, Thermo Scientific Chemicals
Ammoniumdihydrogenphosphat, 99.9 %, reinst, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammoniumdihydrogenphosphat, 99.9 %, reinst, Thermo Scientific Chemicals

CAS: 7722-76-1 | H6NO4P | 115.03 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
100 g
500 g
2.5 kg
Verpackung:
Kunststoffflasche
Katalognummer 193705000
Preis (EUR)
82,65
Exklusiv online
91,50
Ersparnis 8,85 (10%)
Each
Zum Warenkorb hinzufügen
Menge:
500 g
Verpackung:
Kunststoffflasche
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
82,65
Exklusiv online
91,50
Ersparnis 8,85 (10%)
Each
Zum Warenkorb hinzufügen
Chemikalien-Kennzeichnungen
CAS7722-76-1
IUPAC Namephosphoric acid amine
Molecular FormulaH6NO4P
InChI KeyLFVGISIMTYGQHF-UHFFFAOYSA-N
SMILESN.OP(O)(O)=O
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Total trace metal impurities=<0.1 %
Appearance (Color)White
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only