Zinc oxide, 99.5+%
Zinc oxide, 99.5+%
Zinc oxide, 99.5+%
Thermo Scientific Chemicals

Zinc oxide, 99.5+%

CAS: 1314-13-2 | OZn | 81.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Packaging:
View all
Plastic Drum
Catalog number 194890050
Price (USD)
160.65
Online Exclusive
178.00
Save 17.35 (10%)
Each
Quantity:
5 kg
Packaging:
Plastic Drum
Request bulk or custom format
Price (USD)
160.65
Online Exclusive
178.00
Save 17.35 (10%)
Each
Chemical Identifiers
CAS1314-13-2
IUPAC Nameoxozinc
Molecular FormulaOZn
InChI KeyXLOMVQKBTHCTTD-UHFFFAOYSA-N
SMILESO=[Zn]
View more
SpecificationsSpecification SheetSpecification Sheet
Carbonate+subst.insol.in acidsPasses test
Carbonate+subst.insol.in acidsPasses test
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only