Thermo Scientific Chemicals

Ammonium metavanadate, 99.5%, for analysis, Thermo Scientific Chemicals

Catalog number: 194910050
5 kg, Plastic Drum, Each
Thermo Scientific Chemicals

Ammonium metavanadate, 99.5%, for analysis, Thermo Scientific Chemicals

Catalog number: 194910050
5 kg, Plastic Drum, Each
Quantity
Packaging
Catalog number: 194910050
Price (USD)
Quantity
-

Chemical Identifiers

CAS
7803-55-6
IUPAC Name
vanadiumoylol amine
Molecular Formula
H4NO3V
InChI Key
YBVKNHXQSRDWAA-UHFFFAOYSA-M
SMILES
N.O[V](=O)=O
Copper (Cu)
=<10 ppm
Appearance (Form)
Crystalline powder
Cobalt (Co)
=<20 ppm
Lead (Pb)
=<20 ppm
Iron (Fe)
=<10 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text