Potassium hexachloroplatinate(IV), ca. 40% Pt
Potassium hexachloroplatinate(IV), ca. 40% Pt
Potassium hexachloroplatinate(IV), ca. 40% Pt
Thermo Scientific Chemicals

Potassium hexachloroplatinate(IV), ca. 40% Pt

CAS: 16921-30-5 | Cl6K2Pt | 485.98 g/mol
数量:
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 195350250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS16921-30-5
IUPAC Namedipotassium hexachloroplatinumdiuide
Molecular FormulaCl6K2Pt
InChI KeyDPAIVKJGTXERIM-UHFFFAOYSA-H
SMILES[K+].[K+].Cl[Pt--](Cl)(Cl)(Cl)(Cl)Cl
さらに表示
Appearance (Form)Powder and/or chunks
Appearance (Color)Yellow to light orange
Platinum (Pt)39.4 to 40.6 % (Pt)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only