Platinum(II) bromide, 98%
Platinum(II) bromide, 98%
Platinum(II) bromide, 98%
Thermo Scientific Chemicals

Platinum(II) bromide, 98%

CAS: 13455-12-4 | Br2Pt | 354.89 g/mol
Have Questions?
Catalog NumberQuantityPackaging
1954100101 gGlass bottle
Catalog number 195410010
Price (USD)
136.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
136.00
Each
Chemical Identifiers
CAS13455-12-4
IUPAC Nameplatinum(2+) dibromide
Molecular FormulaBr2Pt
InChI KeyKGRJUMGAEQQVFK-UHFFFAOYSA-L
SMILES[Br-].[Br-].[Pt++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown to khaki
Assay>=97.5 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only