Ruthenium(III) chloride hydrate, 35 - 40% Ru
Ruthenium(III) chloride hydrate, 35 - 40% Ru
Ruthenium(III) chloride hydrate, 35 - 40% Ru
Thermo Scientific Chemicals

Ruthenium(III) chloride hydrate, 35 - 40% Ru

CAS: 14898-67-0 | Cl3Ru | 207.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
100 g
Packaging:
Glass bottle
Catalog number 195481000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS14898-67-0
IUPAC Nameruthenium(3+) trichloride
Molecular FormulaCl3Ru
InChI KeyYBCAZPLXEGKKFM-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Ru+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Platinum (Pt)=<500 ppm
Rhodium (Rh)=<500 ppm
Silicon (Si)=<300 ppm
Appearance (Color)Dark brown to black
Appearance (Form)Powder or crystalline powder or crystals and/or
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only