Potassium dichromate, 99.5%, for analysis
Potassium dichromate, 99.5%, for analysis
Potassium dichromate, 99.5%, for analysis
Potassium dichromate, 99.5%, for analysis
Potassium dichromate, 99.5%, for analysis
Thermo Scientific Chemicals

Potassium dichromate, 99.5%, for analysis

CAS: 7778-50-9 | Cr2K2O7 | 294.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
500 g
2 kg
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 196595000
Price (EUR)
112,00
Each
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
112,00
Each
Add to cart
Chemical Identifiers
CAS7778-50-9
IUPAC Namedipotassium [(oxidodioxochromio)oxy]chromiumoylolate
Molecular FormulaCr2K2O7
InChI KeyKMUONIBRACKNSN-UHFFFAOYSA-N
SMILES[K+].[K+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange
Appearance (Form)Crystalline powder or crystals
Titration Iodometric>=99.5 %
Loss on drying=<0.05 % (105°C)
Chloride (Cl)=<10 ppm
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Oxidation reagent
RUO – Research Use Only