3-Pentyn-1-ol, 99%
3-Pentyn-1-ol, 99%
3-Pentyn-1-ol, 99%
Thermo Scientific Chemicals

3-Pentyn-1-ol, 99%

CAS: 10229-10-4 | C5H8O | 84.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 197331000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS10229-10-4
IUPAC Namepent-3-yn-1-ol
Molecular FormulaC5H8O
InChI KeyIDYNOORNKYEHHO-UHFFFAOYSA-N
SMILESCC#CCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear yellow
Infrared spectrumConforms
Refractive index1.4550 to 1.4570 (20°C, 589 nm)
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only