n-Hexane, 95+%, extra pure
n-Hexane, 95+%, extra pure
n-Hexane, 95+%, extra pure
Thermo Scientific Chemicals

n-Hexane, 95+%, extra pure

CAS: 110-54-3 | C6H14 | 86.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
5 L
25 L
Packaging:
View all
Metal Drum
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 197360250
Price (USD)
-
Quantity:
25 L
Packaging:
Metal Drum
Request bulk or custom format
Chemical Identifiers
CAS110-54-3
IUPAC Namehexane
Molecular FormulaC6H14
InChI KeyVLKZOEOYAKHREP-UHFFFAOYSA-N
SMILESCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<20 APHA
Residue after evaporation=<0.005 %
Water=<0.05 % (K.F.)
Appearance (Form)Clear liquid
GC>=95.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only