Lead(II) oxide, 99+%
Lead(II) oxide, 99+%
Lead(II) oxide, 99+%
Thermo Scientific Chemicals

Lead(II) oxide, 99+%

CAS: 1317-36-8 | OPb | 223.20 g/mol
Have Questions?
Catalog NumberQuantityPackaging
1993400101 kgGlass bottle
Catalog number 199340010
Price (USD)
80.20
Each
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
80.20
Each
Chemical Identifiers
CAS1317-36-8
IUPAC Nameplumbanone
Molecular FormulaOPb
InChI KeyYEXPOXQUZXUXJW-UHFFFAOYSA-N
SMILESO=[Pb]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to light orange
Appearance (Form)Powder
Titration Complexometric99.0 to 101.0 %
Loss on drying=<0.1 % (105°C)
Silver (Ag)=<50 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only