Lithium aluminum hydride, 4.0M solution in diethyl ether, AcroSeal™
Lithium aluminum hydride, 4.0M solution in diethyl ether, AcroSeal™
Lithium aluminum hydride, 4.0M solution in diethyl ether, AcroSeal™
Thermo Scientific Chemicals

Lithium aluminum hydride, 4.0M solution in diethyl ether, AcroSeal™

CAS: 16853-85-3 | AlH4Li | 37.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
View all
AcroSeal™ Glass Bottle
Catalog number 199518000
Price (EUR)
324,00
Each
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (EUR)
324,00
Each
Chemical Identifiers
CAS16853-85-3
IUPAC Namelithium(1+) alumanuide
Molecular FormulaAlH4Li
InChI KeyOCZDCIYGECBNKL-UHFFFAOYSA-N
SMILES[Li+].[AlH4-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
AppearanceMay develop some turbidity or precipitate
Titration Iodometric18.0 to 22.0 % (LiAlH4)
Appearance (Color)Colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only