Diisobutylaluminium hydride, 1.1M solution in cyclohexane, AcroSeal™
Diisobutylaluminium hydride, 1.1M solution in cyclohexane, AcroSeal™
Diisobutylaluminium hydride, 1.1M solution in cyclohexane, AcroSeal™
Thermo Scientific Chemicals

Diisobutylaluminium hydride, 1.1M solution in cyclohexane, AcroSeal™

CAS: 1191-15-7 | C8H19Al
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
View all
AcroSeal™ Glass bottle
Catalog number 201031000
Price (USD)
65.65
Special offer
Online exclusive
Ends: 28-Dec-2026
77.20
Save 11.55
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (USD)
65.65
Special offer
Online exclusive
Ends: 28-Dec-2026
77.20
Save 11.55
Each
Chemical Identifiers
CAS1191-15-7
IUPAC Namebis(2-methylpropyl)alumanyl
Molecular FormulaC8H18Al
InChI KeySIPUZPBQZHNSDW-UHFFFAOYSA-N
SMILESCC(C)C[Al]CC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Strength19.5 to 20.5 % (DIBAL-H, on Al basis)
Appearance (Form)Clear solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid, Reduction reagent
RUO – Research Use Only