Rubidium chloride, 99.8+%, (trace metal basis)
Rubidium chloride, 99.8+%, (trace metal basis)
Rubidium chloride, 99.8+%, (trace metal basis)
Thermo Scientific Chemicals

Rubidium chloride, 99.8+%, (trace metal basis)

CAS: 7791-11-9 | ClRb | 120.92 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Catalog number 201260500
Price (USD)
249.65
Special offer
Online exclusive
Ends: 28-Dec-2026
294.00
Save 44.35
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
249.65
Special offer
Online exclusive
Ends: 28-Dec-2026
294.00
Save 44.35
Each
Chemical Identifiers
CAS7791-11-9
IUPAC Namerubidium(1+) chloride
Molecular FormulaClRb
InChI KeyFGDZQCVHDSGLHJ-UHFFFAOYSA-M
SMILES[Cl-].[Rb+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals and/or chunks
Total trace metal impurities=<2000.0 ppm
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only