Tetrabutylammonium fluoride, 1M solution in THF, containing ca. 5% water
Tetrabutylammonium fluoride, 1M solution in THF, containing ca. 5% water
Tetrabutylammonium fluoride, 1M solution in THF, containing ca. 5% water
Thermo Scientific Chemicals

Tetrabutylammonium fluoride, 1M solution in THF, containing ca. 5% water

CAS: 429-41-4 | C16H36FN | 261.47 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
100 mL
500 mL
2.5 L
Catalog number 201950050
Price (USD)
-
Quantity:
5 mL
Request bulk or custom format
Chemical Identifiers
CAS429-41-4
IUPAC Nametetrabutylazanium fluoride
Molecular FormulaC16H36FN
InChI KeyFPGGTKZVZWFYPV-UHFFFAOYSA-M
SMILES[F-].CCCC[N+](CCCC)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Appearance (Color)Clear yellow to amber to light green to brown
Water=<8 %
Titration with HClO40.90 to 1.10 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only