Amberlite™ IRA-410, Thermo Scientific Chemicals
Amberlite™ IRA-410, Thermo Scientific Chemicals
Amberlite™ IRA-410, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Amberlite™ IRA-410, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantityPackaging
202295000500 gPlastic bottle
Catalog number 202295000
Price (USD)
47.65
Online Exclusive
52.60
Save 4.95 (9%)
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
47.65
Online Exclusive
52.60
Save 4.95 (9%)
Each
Chemical Identifiers
CAS9002-26-0
MDL NumberMFCD00132710
SpecificationsSpecification SheetSpecification Sheet
Uniformity coefficient=<1.60
Total volume capacity>=1.25 eq/l (Cl-)
Particle size=<1.0 % (<0.300 mm)
Appearance (Color)Transparent light yellow to light orange
Moisture holding capacity45.0 to 51.0 % (Cl-)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ion exchange resin
RUO – Research Use Only