Tetrakis(triphenylphosphine)palladium(0), 99%
Tetrakis(triphenylphosphine)palladium(0), 99%
Tetrakis(triphenylphosphine)palladium(0), 99%
Thermo Scientific Chemicals

Tetrakis(triphenylphosphine)palladium(0), 99%

CAS: 14221-01-3 | C72H60P4Pd | 1155.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 202380010
Price (USD)
62.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
62.00
Each
Chemical Identifiers
CAS14221-01-3
IUPAC Nametetrakis(triphenylphosphane) palladium
Molecular FormulaC72H60P4Pd
InChI KeyNFHFRUOZVGFOOS-UHFFFAOYSA-N
SMILES[Pd].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to yellow-green
Infrared spectrumConforms
Carbon (C)73.7 to 76.0 %
Appearance (Form)Powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only