6-Aminonicotinic acid, 98%
6-Aminonicotinic acid, 98%
6-Aminonicotinic acid, 98%
Thermo Scientific Chemicals

6-Aminonicotinic acid, 98%

CAS: 3167-49-5 | C6H6N2O2 | 138.13 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
產品號碼 202540050
價格 (TWD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS3167-49-5
IUPAC Name6-aminopyridine-3-carboxylic acid
Molecular FormulaC6H6N2O2
InChI KeyZCIFWRHIEBXBOY-UHFFFAOYSA-N
SMILESNC1=CC=C(C=N1)C(O)=O
檢視更多
規格Specification Sheet規格表
Appearance (Form)Powder
Titration with HClO4>=97.5 %
Appearance (Color)White to beige
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only