D(-)-Gulonic acid gamma-lactone, 99%
D(-)-Gulonic acid gamma-lactone, 99%
D(-)-Gulonic acid gamma-lactone, 99%
D(-)-Gulonic acid gamma-lactone, 99%
Thermo Scientific Chemicals

D(-)-Gulonic acid gamma-lactone, 99%

CAS: 6322-07-2 | C6H10O6 | 178.14 g/mol
Have Questions?
Catalog NumberQuantityPackaging
20445025025 gGlass bottle
Catalog number 204450250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6322-07-2
IUPAC Name5-(1,2-dihydroxyethyl)-3,4-dihydroxyoxolan-2-one
Molecular FormulaC6H10O6
InChI KeySXZYCXMUPBBULW-UHFFFAOYNA-N
SMILESOCC(O)C1OC(=O)C(O)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Appearance (Color)White
Infrared spectrumConforms
Loss on drying=<1 %
HPLC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only