Magnesium oxide, 98%, pure
Magnesium oxide, 98%, pure
Magnesium oxide, 98%, pure
Thermo Scientific Chemicals

Magnesium oxide, 98%, pure

CAS: 1309-48-4 | MgO | 40.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
View all
Plastic Bottle
Catalog number 205150025
Price (USD)
194.00
Each
Quantity:
2.5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
194.00
Each
Chemical Identifiers
CAS1309-48-4
IUPAC Nameoxomagnesium
Molecular FormulaMgO
InChI KeyCPLXHLVBOLITMK-UHFFFAOYSA-N
SMILESO=[Mg]
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Complexometric>=97.5 %
Appearance (Form)Crystalline powder
Appearance (Color)White to grey
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only