Cesium chloride, 99.99%, (trace metal basis), for spectroscopy, optical grade
Cesium chloride, 99.99%, (trace metal basis), for spectroscopy, optical grade
Cesium chloride, 99.99%, (trace metal basis), for spectroscopy, optical grade
Thermo Scientific Chemicals

Cesium chloride, 99.99%, (trace metal basis), for spectroscopy, optical grade

CAS: 7647-17-8 | ClCs | 168.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 206320250
Price (TWD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7647-17-8
IUPAC Namecaesium(1+) chloride
Molecular FormulaClCs
InChI KeyAIYUHDOJVYHVIT-UHFFFAOYSA-M
SMILES[Cl-].[Cs+]
View more
SpecificationsSpecification SheetSpecification Sheet
Insoluble matter=<25 ppm (in water)
UVat 280 nm A: =<0.02
Appearance (Form)Crystalline powder or crystals
Purity>=99.94 % (metals basis)
UV(50 % in water) 1 cm cell
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis, For UV/VIS spectroscopy
RUO – Research Use Only