Sodium phosphate, tribasic dodecahydrate, 98%, for analysis
Sodium phosphate, tribasic dodecahydrate, 98%, for analysis
Sodium phosphate, tribasic dodecahydrate, 98%, for analysis
Thermo Scientific Chemicals

Sodium phosphate, tribasic dodecahydrate, 98%, for analysis

CAS: 10101-89-0 | Na3O4P·12H2O | 380.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
25 kg
Packaging:
View all
Plastic bottle
Catalog number 206520010
Price (USD)
64.65
Special offer
Online exclusive
Ends: 28-Dec-2026
75.80
Save 11.15
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
64.65
Special offer
Online exclusive
Ends: 28-Dec-2026
75.80
Save 11.15
Each
Chemical Identifiers
CAS10101-89-0
IUPAC Nametrisodium dodecahydrate phosphate
Molecular FormulaH24Na3O16P
InChI KeyASTWEMOBIXQPPV-UHFFFAOYSA-K
SMILESO.O.O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[O-]P([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Chloride (Cl)=<100 ppm
Appearance (Color)Colorless to white
Titration excess HCl97.5 to 102.5 %
Heavy metals (as Pb)=<20 ppm
Iron (Fe)=<50 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only