Is Dimethyl sulfoxide (DMSO) (Cat. No. 20688 and 85190) suitable for cell freezing?
The DMSO with Cat. Nos. 20688 and 85190 have not been tested for cell freezing. However, Dimethyl sulfoxide, Bioreagent, Thermo Scientific Chemicals (Cat. No. J66650-AE, J66650-AK, J66650-AD) is ⋝99.9% pure and can be used for cell freezing.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center