Silver(II) oxide, 85.5-88.0% Ag
Silver(II) oxide, 85.5-88.0% Ag
Silver(II) oxide, 85.5-88.0% Ag
Thermo Scientific Chemicals

Silver(II) oxide, 85.5-88.0% Ag

CAS: 1301-96-8 | AgO | 123.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 207850100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1301-96-8
IUPAC Nameoxosilver
Molecular FormulaAgO
InChI KeyOTCVAHKKMMUFAY-UHFFFAOYSA-N
SMILESO=[Ag]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey to black
Appearance (Form)Powder
Assay85.5 to 88 % (AG)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only